Learn More
SCAT HPLC Safety Waste Caps, V2.0
SCAT Safety Waste Caps protect staff and the environment from harmful solvent vapours. With a suitable exhaust filter, they turn your disposal container into a closed system without escaping solvent vapours.
1135.00 NOK - 2285.00 NOK
Specifications
Type | Safety Waste Cap |
---|---|
For Use With (Application) | Waste Disposal System |
For Use With (Equipment) | Waste Disposal Container |
Product Code | Brand | Thread Size | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Thread Size | Includes | Price | Quantity & Availability | |||||
17122576
![]() |
SCAT
307108 |
S40/GL40 | 3 x 2.3/3.2 mm OD + connector for exhaust filter |
1280.00 NOK
Each |
Estimated Shipment: 31-07-2025
Log in to see stock. |
|||||
16443809
![]() |
SCAT
307923 |
GL 45 | 2 x PFA-fitting 2.3/3.2 mm OD, 2 x PFA-blind plug, 1 x 5.0 - 11.5 mm ID + connector for exhaust filter |
1205.00 NOK
Each |
Estimated Shipment: 21-07-2025
Log in to see stock. |
|||||
16413809
![]() |
SCAT
307912 |
GL 45 | 3 x PFA-fitting 2.3/3.2 mm OD, 1 x PFA-blind plug + connector for exhaust filter |
1135.00 NOK
Each |
Estimated Shipment: 29-07-2025
Log in to see stock. |
|||||
16439963
![]() |
SCAT
308921 |
GL 45 | 4 x PFA-fitting 1.6/2.3/3.2 mm OD, 4 x PFA-blind plug, 1 x 5.0 - 11.5 mm ID, 1 x PTFE-blind plug + connector for exhaust filter |
1690.00 NOK
Each |
Estimated Shipment: 29-07-2025
Log in to see stock. |
|||||
16526034
![]() |
SCAT
307925 |
S60/61 | 2 x 2.3/3.2 mm OD, 1 x 5.0 - 11.5 mm ID + connector for exhaust filter |
1550.00 NOK
Each |
Estimated Shipment: 24-07-2025
Log in to see stock. |
|||||
16635722
![]() |
SCAT
307500 |
S60 | 3 x 1.6/2.3/3.2 mm OD, 3 x PFA-blind plug, 3 x 5.0 - 11.5 mm ID, 3 x PTFE-blind plug + connector for exhaust filter |
2095.00 NOK
Each |
Estimated Shipment: 29-07-2025
Log in to see stock. |
|||||
16626214
![]() |
SCAT
307931 |
S60/61 | 4 x 2.3/3.2 mm OD, 1 x 5.0 - 11.5 mm ID + connector for exhaust filter |
1975.00 NOK
Each |
Estimated Shipment: 31-07-2025
Log in to see stock. |
|||||
17089693
![]() |
SCAT
307918 |
S60/61 | 3 x 2.3/3.2 mm OD + connector for exhaust filter |
1550.00 NOK
Each |
Estimated Shipment: 31-07-2025
Log in to see stock. |
|||||
17175501
![]() |
SCAT
307916 |
S60/61 | 3 x 2.3/3.2 mm OD, grounding connection + connector for exhaust filter |
2260.00 NOK
Each |
Estimated Shipment: 05-08-2025
Log in to see stock. |
|||||
18696468
|
SCAT
307970 |
S60/61 | 1x NPT 3/8 in. (f), 1x connection for exhaust filter (45° beveled) |
2285.00 NOK
1 sheet |
Estimated Shipment: 29-07-2025
Log in to see stock. |
|||||
Description
- Discover the flexibility of the world's leading system for solvent safety
- SafetyWasteCap with S 60/61 thread in the new design of generation 2.0
- Optimum protection when collecting hazardous or toxic liquid waste
- Flexible system with fittings and tube connectors in different sizes
- With multiple options to connect different tubing and capillary sizes
- Time saving in changing the waste containers while tubes are fixed
- Resistant against aggressive media
- PTFE Body 360° freely rotatable: Provides easy container change without distortion of tubing
- Body with integrated FKM Seal: Provides hermetical connection to the container opening and adapts smoothly even to rugged glass surfaces. An additional PTFE layer protects the seal from direct contact to solvents.
- PFA Fittings: Improved design for easy tubing connection - Excellent chemical resistance - Flammability classification V-0 (UL-94)
- Medical Grade PTFE: Excellent chemical resistance Food safe FDA compliant -
- Flammability classification V-0 (UL-94)
- Insertion Label: Mark your bottles with individual descriptions
- PPS Screw Cap: Autoclavable / sterilisable up to 200°C - Flammability classification V-0 (UL-94)
- Ergonomic Design: Enhanced stability and construction - Improved handling - Easy installation - Smart exchange
- Huge range of accessories, optional: exhaust filters, funnel use, level control preventing overflowing containers
Specifications
Safety Waste Cap | |
Waste Disposal System | |
Waste Disposal Container |
Product Title
SCAT HPLC Safety Waste Caps, V2.0