Learn More
SymLine™ Safety Waste Caps
These Safety Waste Caps safely connect the pipework to the waste container as an integrated exhaust filter blocks solvent vapors for health and environmental protection.
Brand: SymLine™ 307971
This item is not returnable.
View return policy
Description
- Available with different thread sizes for attachment to pipe system
- Different numbers of hose connections for disposal of liquid waste
- Available with electronic level sensors that indicate the degree of filling of the disposal container
- By using a 4-in-1 collector (306509), up to 4 Safety Waste Caps can be connected simultaneously to only one connection point on the pipe system
Specifications
Waste Disposal System | |
1x NPT 3/8 in. (f), 1x connection for exhaust filter (45° beveled) | |
S60/61 |
Waste Disposal Container | |
PTFE | |
Safety Waste Collector |