Learn More
SCAT™ Safety Waste Cap 'LISA', V3.0
The latest generation of a modern and universally expandable modular system for the safe disposal of liquid HPLC or UHPLC waste.
1480.00 NOK - 2445.00 NOK
Specifications
Type Waste Cap
For Use With (Application) HPLC and UHPLC Systems
View More Specs
Products 20
Product Code Brand Thread Size Includes Price Quantity & Availability  
Product Code Brand Thread Size Includes Price Quantity & Availability  
17285044
View Documents Promotion Details
SCAT™
350045
G45 4 Fittings (1.6/2.3/3.2 mm OD), 4 PFA Blind Plugs, 3 Fittings (5 to 11.5 mm ID), 3 PTFE Blind Plug, Blind Plug for Exhaust Filter
2260.00 NOK
Each
Only null left
Estimated Shipment: 09-07-2025
Add to basket
17295044
View Documents
SCAT™
350050
S50 4 Fittings (1.6/2.3/3.2 mm OD), 4 PFA Blind Plugs, 3 Fittings (5 to 11.5 mm ID), 3 PTFE Blind Plug, Blind Plug for Exhaust Filter
2190.00 NOK
Each
Only null left
Estimated Shipment: 02-07-2025
Add to basket
17878733
View Documents
SCAT™
450050
S50 4 Fittings (1.6/2.3/3.2 mm OD), 4 PFA Blind Plugs, 3 Fittings (5 to 11.5 mm ID), 3 PTFE Blind Plugs, Connector for Exhaust Filter
2380.00 NOK
Each
Only null left
Estimated Shipment: 07-07-2025
Add to basket
17205054
View Documents
SCAT™
350051
S51 4 Fittings (1.6/2.3/3.2 mm OD), 4 PFA Blind Plugs, 3 Fittings (5 to 11.5 mm ID), 3 PTFE Blind Plug, Blind Plug for Exhaust Filter
2190.00 NOK
Each
Only null left
Estimated Shipment: 02-07-2025
Add to basket
17225054
View Documents Promotion Details
SCAT™
350055
S55 4 Fittings (1.6/2.3/3.2 mm OD), 4 PFA Blind Plugs, 3 Fittings (5 to 11.5 mm ID), 3 PTFE Blind Plug, Blind Plug for Exhaust Filter
2260.00 NOK
Each
Only null left
Estimated Shipment: 02-07-2025
Add to basket
17898733
View Documents
SCAT™
450055
S55 4 Fittings (1.6/2.3/3.2 mm OD), 4 PFA Blind Plugs, 3 Fittings (5 to 11.5 mm ID), 3 PTFE Blind Plugs, Connector for Exhaust Filter
2380.00 NOK
Each
Only null left
Estimated Shipment: 07-07-2025
Add to basket
17235054
View Documents Promotion Details
SCAT™
350060
S60/61 4 Fittings (1.6/2.3/3.2 mm OD), 4 PFA Blind Plugs, 3 Fittings (5 to 11.5 mm ID), 3 PTFE Blind Plug, Blind Plug for Exhaust Filter
2050.00 NOK
Each
Only null left
Estimated Shipment: 08-07-2025
Add to basket
17225064
View Documents
SCAT™
450060
S60/61 4 Fittings (1.6/2.3/3.2 mm OD), 4 PFA Blind Plugs, 3 Fittings (5 to 11.5 mm ID), 3 PTFE Blind Plug, Blind Plug for Exhaust Filter
2445.00 NOK
Each
Only null left
Estimated Shipment: 02-07-2025
Add to basket
17255054
View Documents
SCAT™
350065
S65 4 Fittings (1.6/2.3/3.2 mm OD), 4 PFA Blind Plugs, 3 Fittings (5 to 11.5 mm ID), 3 PTFE Blind Plug, Blind Plug for Exhaust Filter
2190.00 NOK
Each
Only null left
Estimated Shipment: 02-07-2025
Add to basket
17818743
View Documents
SCAT™
450070
S70 4 Fittings (1.6/2.3/3.2 mm OD), 4 PFA Blind Plugs, 3 Fittings (5 to 11.5 mm ID), 3 PTFE Blind Plugs, Connector for Exhaust Filter
2380.00 NOK
Each
Only null left
Estimated Shipment: 07-07-2025
Add to basket
17265054
View Documents
SCAT™
350070
S70/71 4 Fittings (1.6/2.3/3.2 mm OD), 4 PFA Blind Plugs, 3 Fittings (5 to 11.5 mm ID), 3 PTFE Blind Plug, Blind Plug for Exhaust Filter
2190.00 NOK
Each
Only null left
Estimated Shipment: 02-07-2025
Add to basket
17285054
View Documents
SCAT™
350090
S90 4 Fittings (1.6/2.3/3.2 mm OD), 4 PFA Blind Plugs, 3 Fittings (5 to 11.5 mm ID), 3 PTFE Blind Plug, Blind Plug for Exhaust Filter
2190.00 NOK
Each
Only null left
Estimated Shipment: 02-07-2025
Add to basket
17295054
View Documents
SCAT™
350095
S95 4 Fittings (1.6/2.3/3.2 mm OD), 4 PFA Blind Plugs, 3 Fittings (5 to 11.5 mm ID), 3 PTFE Blind Plug, Blind Plug for Exhaust Filter
2190.00 NOK
Each
Only null left
Estimated Shipment: 02-07-2025
Add to basket
17215064
View Documents Promotion Details
SCAT™
450045
GL45 4 Fittings (1.6/2.3/3.2 mm OD), 4 PFA Blind Plugs, 3 Fittings (5 to 11.5 mm ID), 3 PTFE Blind Plug, Blind Plug for Exhaust Filter
2445.00 NOK
Each
Only null left
Estimated Shipment: 02-07-2025
Add to basket
17205064
View Documents
SCAT™
350100
GL45 4 Fittings (1.6/2.3/3.2 mm OD), 4 PFA Blind Plugs, 3 Fittings (5 to 11.5 mm ID), 3 PTFE Blind Plug, Blind Plug for Exhaust Filter
1480.00 NOK
Each
Only null left
Estimated Shipment: 02-07-2025
Add to basket
17235064
View Documents
SCAT™
450100
GL45 4 Fittings (1.6/2.3/3.2 mm OD), 4 PFA Blind Plugs, 3 Fittings (5 to 11.5 mm ID), 3 PTFE Blind Plug, Blind Plug for Exhaust Filter
1765.00 NOK
Each
Only null left
Estimated Shipment: 02-07-2025
Add to basket
17215054
View Documents
SCAT™
350053
B53 4 Fittings (1.6/2.3/3.2 mm OD), 4 PFA Blind Plugs, 3 Fittings (5 to 11.5 mm ID), 3 PTFE Blind Plug, Blind Plug for Exhaust Filter
2190.00 NOK
Each
Only null left
Estimated Shipment: 02-07-2025
Add to basket
17245054
View Documents
SCAT™
350063
B63 4 Fittings (1.6/2.3/3.2 mm OD), 4 PFA Blind Plugs, 3 Fittings (5 to 11.5 mm ID), 3 PTFE Blind Plug, Blind Plug for Exhaust Filter
2190.00 NOK
Each
Only null left
Estimated Shipment: 02-07-2025
Add to basket
17275054
View Documents
SCAT™
350083
B83 4 Fittings (1.6/2.3/3.2 mm OD), 4 PFA Blind Plugs, 3 Fittings (5 to 11.5 mm ID), 3 PTFE Blind Plug, Blind Plug for Exhaust Filter
2190.00 NOK
Each
Only null left
Estimated Shipment: 02-07-2025
Add to basket
17885724
View Documents
SCAT™
450090
S90 4 x 1.6/2.3/3.2 mm OD, 4 x PFA-blind plug, 3 x 5 to 11.5 mm ID, 3 x PTFE-blind plug + connector for exhaust filter
2415.00 NOK
1 set
Only null left
Estimated Shipment: 04-07-2025
Add to basket
Description
Description
The basic module includes a Safety Waste Cap V3.0 'LISA' with your selected thread size. Included in the scope of delivery are a further 23 accessories. In addition, a variety of other SCAT™ accessories such as a funnel or an exhaust air filter can be docked to the Safety Waste Cap LISA V3.0. You will find a selection listed in the ordering table:
  • Satelite blind plug for extending and attaching various components such as a funnel
  • Tube connector for connection for larger tubes with 5 to 11.5 mm ID
  • Satellite made of PTFE for optimal chemical resistance
  • 360° freely rotatable screw cap for easy changing of the container without twisting the tubing
  • Fittings with improved design for easier connection of capillaries with 1.6 mm, 2.3 mm or 3.2 mm OD
  • SCAT™ exhaust filters block harmful vapors with the help of optimized activated carbon and ensure safe pressure equalization in the waste containers
  • Huge scope of delivery of fittings, blind plugs, and accessories ensures that docking system is ready for immediate use
Specifications
Specifications
Waste Cap
HPLC and UHPLC Systems
Documents
Product Certifications
Product Content Correction
Product Title
SCAT™ Safety Waste Cap 'LISA', V3.0