Learn More
Brady™ Safety Sign COVID-19 Keep Social Distance
An efficient solution with clear safety instructions to easily communicate COVID-19 countermeasures on work floors.
138.00 NOK - 163.00 NOK
Specifications
Material | Laminated Polyester |
---|---|
Mounting Type | Self-adhesive |
Dimensions (L x W) | 800 x 80 mm (W x H) |
Product Code | Brand | Languages | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Languages | Price | Quantity & Availability | ||||||
16506424
|
Brady™
306829 |
Dutch |
138.00 NOK
Each |
Estimated Shipment: 12-05-2025
Log in to see stock. |
||||||
16506444
|
Brady™
306849 |
Swedish |
160.00 NOK
Each |
Estimated Shipment: 15-05-2025
Log in to see stock. |
||||||
16516424
|
Brady™
306830 |
French |
163.00 NOK
Each |
Estimated Shipment: 12-05-2025
Log in to see stock. |
||||||
16526424
|
Brady™
306831 |
German |
163.00 NOK
Each |
Estimated Shipment: 15-05-2025
Log in to see stock. |
||||||
16536424
|
Brady™
306832 |
English |
163.00 NOK
Each |
Estimated Shipment: 11-06-2025
Log in to see stock. |
||||||
16546424
|
Brady™
306833 |
Spanish |
160.00 NOK
Each |
Estimated Shipment: 12-05-2025
Log in to see stock. |
||||||
16556424
|
Brady™
306834 |
Italian |
160.00 NOK
Each |
Estimated Shipment: 15-05-2025
Log in to see stock. |
||||||
16556444
|
Brady™
306854 |
Norwegian | N/A | N/A | N/A | |||||
16586434
|
Brady™
306847 |
Danish |
160.00 NOK
Each |
Estimated Shipment: 15-05-2025
Log in to see stock. |
||||||
16596434
|
Brady™
306848 |
Finnish |
160.00 NOK
Each |
Estimated Shipment: 15-05-2025
Log in to see stock. |
||||||
Description
- Provides clear, visual communication where it matters
- Highlights the use of appropriate personal protection
- Easily communicates social distancing and hygiene
Specifications
Laminated Polyester | |
Self-adhesive | |
800 x 80 mm (W x H) |
Product Title
Brady™ Safety Sign COVID-19 Keep Social Distance