Description
- Provide protection against liquid splash and dust
- Indirect ventilation design
- Strong polycarbonate lens for impact resistance
Specifications
Specifications
Lens Tint | Clear |
Product Type | Safety Goggles |
Ventilation Details | Indirect vented |
Lens Material | Polycarbonate |
Special Offers
-
Multiple Manufacturers
Gear Up for Safety
Save up to 60% on gloves, apparel, and morePromo Code: 24131Expires: 2025-06-08
New Offer Shop Now
Product Content Correction
Product Title
3M™ Safety Goggles 71347-00011 >