Learn More
3M™ Safety Glasses Carrying Case
Click to view available options
Style:
Rigid
Semi Rigid
Soft with Zipper
Unit Size:
Each
Pack of 20
Description
- Helps protect your 3M™ Safety Glasses and improves lifetime.
- Choose from several styles:
- Rigid safety glasses carrying case features a snap clasp and belt loop that allows you to easily carry your glasses.
- Semi-rigid safety glasses carrying case with belt clip for attaching to belt loops.
- Soft safety glasses carrying case features a zipper, belt loop, and snap clasp.
Specifications
Specifications
Description | Rigid |
Color | Black |
Product Type | Eyewear Case |
For Use With (Equipment) | Safety Glasses |
Includes | Belt loop |
Model | 12-0200-00 |
Size | Large |
Style | Rigid |
Product Content Correction
Product Title
3M™ Safety Glasses Carrying Case >