Learn More
Bollé Safety Faceshield
Click to view available options
Model:
B-Line
Clear
Flip Up
Tilting
Unit Size:
Each
Description
- Adjustable
Specifications
Specifications
Lens Tint | Clear |
Product Type | Faceshield |
Frame Color | Black |
Includes | Protective bag |
Model | Clear |
No. per Pack | 1 Ea. |
Material | Polycarbonate |
Frame Material | Polypropylene |
Documents
Product Information
Specification Sheets
SPHERE
File size: 605.8 kB
Promotions
-
Multiple Manufacturers
Essential Eye Protection
Your vision deserves the best protection.Promo Code: 24152Expires: 2025-07-04
-
Multiple Manufacturers
Spotlight
Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.Promo Code: 24156Expires: 2025-06-30
Product Content Correction
Product Title
Bollé Safety Faceshield >