Learn More
DuPont™ Tyvek 500 Xpert Hooded Coverall
GREENER_CHOICE
Composed of flash spun high density polyethylene. DuPont™ Tyvek™ Classic Xpert Hooded Coverall is permeable to both air and water vapor, yet repels water-based liquids and aerosols and offers an excellent barrier against fine particles and fibers.
136.00 NOK - 14305.00 NOK
Specifications
Color White
Seam Type Stitched
View More Specs
Products 18
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
11743103
GREENER_CHOICE
View accessory products View Documents
DuPont™
D14663953
Small
11275.00 NOK
Pack of 100
Only null left
Estimated Shipment: 01-08-2025
Add to basket
12971345
GREENER_CHOICE
View accessory products View Documents
DuPont™
D14663953
Small
3020.00 NOK
Pack of 25
Only null left
Estimated Shipment: 06-08-2025
Add to basket
11703103
GREENER_CHOICE
View accessory products View Documents
DuPont™
D14663967
Medium
11275.00 NOK
Pack of 100
Only null left
Estimated Shipment: 01-08-2025
Add to basket
12991345
GREENER_CHOICE
View accessory products View Documents
DuPont™
D14663967
Medium
3020.00 NOK
Pack of 25
Only null left
Estimated Shipment: 30-07-2025
Add to basket
11713103
GREENER_CHOICE
View accessory products missing translation for 'viewDocuments'
DuPont™
D14663977
Large
11275.00 NOK
Pack of 100
Only null left
Estimated Shipment: 23-07-2025
Add to basket
12911355
GREENER_CHOICE
View accessory products View Documents
DuPont™
D14663977
Large
3020.00 NOK
Pack of 25
Only null left
Estimated Shipment: 30-07-2025
Add to basket
11723103
GREENER_CHOICE
View accessory products View Documents
DuPont™
D14663986
X-Large
11275.00 NOK
Pack of 100
Only null left
Estimated Shipment: 30-07-2025
Add to basket
12931355
GREENER_CHOICE
View accessory products View Documents Promotion Details
DuPont™
D14663986
X-Large
3020.00 NOK
Pack of 25
Only null left
Estimated Shipment: 23-07-2025
Add to basket
13025683
GREENER_CHOICE
View accessory products View Documents
DuPont™
D14663986
X-Large
136.00 NOK
Each
Only null left
Estimated Shipment: 23-07-2025
Add to basket
12951355
GREENER_CHOICE
View accessory products View Documents
DuPont™
D14663997
2X-Large
3020.00 NOK
Pack of 25
Only null left
Estimated Shipment: 06-08-2025
Add to basket
11733103
GREENER_CHOICE
View accessory products View Documents
DuPont™
D14663997
2X-Large
11275.00 NOK
Pack of 100
Only null left
Estimated Shipment: 30-07-2025
Add to basket
12961355
GREENER_CHOICE
View accessory products View Documents
DuPont™
D14664003
3X-Large
3195.00 NOK
Pack of 25
Only null left
Estimated Shipment: 30-07-2025
Add to basket
11753103
GREENER_CHOICE
View accessory products View Documents
DuPont™
D14664003
3X-Large
12045.00 NOK
Pack of 100
Only null left
Estimated Shipment: 23-07-2025
Add to basket
13085673
GREENER_CHOICE
View accessory products View Documents
DuPont™
D14664003
3X-Large
146.00 NOK
Each
Only null left
Estimated Shipment: 31-07-2025
Add to basket
17402893
View Documents
DuPont™
D15553613
4X-Large
12560.00 NOK
Pack of 100
Only null left
Estimated Shipment: 22-08-2025
Add to basket
17412893
View Documents
DuPont™
D15553614
5X-Large
13140.00 NOK
Pack of 100
Only null left
Estimated Shipment: 22-08-2025
Add to basket
17422893
View Documents
DuPont™
D15553615
6X-Large
13710.00 NOK
Pack of 100
Only null left
Estimated Shipment: 22-08-2025
Add to basket
17432893
View Documents
DuPont™
D15553616
7X-Large
14305.00 NOK
Pack of 100
Only null left
Estimated Shipment: 22-08-2025
Add to basket
Description
Description
Composed of flash spun high density polyethylene. Dupont™ Tyvek™ Classic Xpert Hooded Coverall is permeable to both air and water vapor, yet repels water-based liquids and aerosols and offers an excellent barrier against fine particles and fibers.
  • Robust yet lightweight (<180g per garment)
  • Elasticated face, wrists and ankles as well as glued-in waist elastic
  • Ample crotch area for freedom of movement
  • Large, easy-to-grasp zipper puller
  • 3-piece hood for optimal fit to head and face when turning
  • White, sizes S to XXXL
Specifications
Specifications
White
Stitched
Documents
Product Certifications
The user should make the risk assessment to determine the protection required.
Product Content Correction
Product Title
DuPont™ Tyvek 500 Xpert Hooded Coverall