Learn More
Tygon S3™ E-3603 Vacuum Tubing
Formulated to resist flex-fatigue and abrasion
7169.87 NOK - 49775.00 NOK
Product Code | Brand | Diameter (English) Inner | Diameter (English) Outer | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (English) Inner | Diameter (English) Outer | Price | Quantity & Availability | |||||
30053203
|
Tygon
50113-70 |
3/4 in. | 1-1/2 in. |
26390.00 NOK
15m |
Estimated Shipment: 12-09-2025
Log in to see stock. |
|||||
30053637
|
Tygon
50113-68 |
1/2 in. | 1-1/8 in. | N/A | ||||||
30053573
|
Tygon
50113-66 |
1/4 in. | 5/8 in. | N/A | ||||||
30053914
|
Tygon
50113-69 |
5/8 in. | 1-3/8 in. | N/A | ||||||
30054222
|
Tygon
50113-67 |
3/8 in. | 7/8 in. | N/A | ||||||
30053333
|
Tygon
50113-71 |
1 in. | 2 in. |
49775.00 NOK
15m |
Estimated Shipment: 12-09-2025
Log in to see stock. |
|||||
30053233
|
Tygon
50113-65 |
3/16 in. | 9/16 in. | N/A | ||||||
Description
Bio-based formulation reduces environmental impact. Lot-to-lot consistency for reproducible results. Easily slips over fittings and grips securely. Contains no BPA or phthalates. Meets all USP Class VI and FDA food contact criteria. . Ideal for general-purpose, laboratory, and single-use applications. Free technical application support available to assist with tubing selectionSpecifications
USP;FDA | |
Tygon S3 E-3603 |
Product Title
Tygon S3™ E-3603 Vacuum Tubing