missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100A11 Mouse anti-Human, Clone: 1A3F1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 60024-1-IG-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100 genes include at least 13 members which are located as a cluster on chromosome 1q21. This protein may function in motility, invasion, and tubulin polymerization. Chromosomal rearrangements and altered expression of this gene have been implicated in tumor metastasis.Specifications
S100A11 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P31949 | |
S100A11 | |
S100A11 Fusion Protein Ag0343 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
1A3F1 | |
Unconjugated | |
S100A11 | |
Calgizzarin, MLN 70, MLN70, Protein S100 A11, Protein S100 C, S100A11, S100C | |
Mouse | |
Protein G | |
RUO | |
6282 | |
-20°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction