Learn More
Thermo Scientific™ S1 Pipet Filler Accessories
GREENER_CHOICE
Incorporate Thermo Scientific™ S1 Pipet Filler Accessories with S1 Pipet Filler.
Brand:  Thermo Scientific™ 9069
172.46 NOK valid until 2025-06-30
Use promo code "24075" to get your promotional price.
 View more versions of this product
Product Code. 11992181
  • 258.00 NOK / Each
The Ultimate Lab Appliances Promotion
Use promo code 24075 at checkout
View Available Promotions
Only null left
In Stock
Description
Complete your S1 Pipet Filler with these accessories and prolong its use.
Features:
  • Wall-mounted holder and table stand offer compact storage between applications. Recharge the pipet filler in both stands by plugging in the charger. Store pipet filler when working in laminar flow cabinets to save space.
  • Wing stand allows user to set the pipet filler down without removing or emptying the serological pipet.
  • Easy to change filter prevents aerosol or liquid cross contamination of the pipet together with autoclavable silicone adapter.
  • 1mL Pipet Support ensures that 1 mL pipets will hold as securely as larger sizes.
Note:
Wing stand not available for use or sale within the United States or for importation into the United States.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
S1 Pipette Filler
Wing Stand
Single Pipet Filler
Certificates
Promotions
Promotions
  • Multiple Manufacturers
    Enjoy discounts up to 78% on top-quality lab equipment and upgrade your laboratory with the best appliances at the lowest prices.
    Promo Code: 24075
    Expires2025-06-30
    Days
    13
    Hours
    9
    Minutes
    17
    Seconds
    17
Product Content Correction
Product Title
Thermo Scientific™ S1 Pipet Filler Accessories > S1 Accessory, Single Pipet Filler Wing Stand; Unit Size: 1 piece