Learn More
Thermo Scientific™ S1 Pipet Filler Accessories
GREENER_CHOICE
Incorporate Thermo Scientific™ S1 Pipet Filler Accessories with S1 Pipet Filler.
Brand:  Thermo Scientific™ 9068
483.10 NOK valid until 2025-06-30
Use promo code "24075" to get your promotional price.
 View more versions of this product
Product Code. 11866244
  • 722.00 NOK / Each
The Ultimate Lab Appliances Promotion
Use promo code 24075 at checkout
View Available Promotions
Only null left
In Stock
Description
Complete your S1 Pipet Filler with these accessories and prolong its use.
Features:
  • Wall-mounted holder and table stand offer compact storage between applications. Recharge the pipet filler in both stands by plugging in the charger. Store pipet filler when working in laminar flow cabinets to save space.
  • Wing stand allows user to set the pipet filler down without removing or emptying the serological pipet.
  • Easy to change filter prevents aerosol or liquid cross contamination of the pipet together with autoclavable silicone adapter.
  • 1mL Pipet Support ensures that 1 mL pipets will hold as securely as larger sizes.
Note:
Wing stand not available for use or sale within the United States or for importation into the United States.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
S1 Pipette Filler
Universal Power Supply
Videos
  • Thermo Scientific - S1 pipette filler - Redefining pipetting performance
    Your time is valuable and faster performance is essential. Trust the S1 pipette filler to deliver everything you need to stay productive in the lab.
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
      Thermo Scientific S1 pipette filler Redefining pipetting performance
    • Thermo Scientific - S1 pipette filler - Redefining pipetting performance
      Your time is valuable and faster performance is essential. Trust the S1 pipette filler to deliver everything you need to stay productive in the lab.
      Video Player is loading.
      Current Time 0:00
      Duration 0:00
      Loaded: 0%
      Stream Type LIVE
      Remaining Time 0:00
      1x
      • Chapters
      • descriptions off, selected
      • captions off, selected
        Thermo Scientific S1 pipette filler Redefining pipetting performance
      Certificates
      Promotions
      Promotions
      • Multiple Manufacturers
        Save up to 59% on some of our most popular products
        Promo Code: 24140
        Expires2025-08-24
        Days
        67
        Hours
        21
        Minutes
        36
        Seconds
        44
      • Multiple Manufacturers
        Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.
        Promo Code: 24156
        Expires2025-06-30
        Days
        12
        Hours
        21
        Minutes
        36
        Seconds
        44
        Hot Deal
        *Discount will apply to qualifying orders received by Fisher Scientific between 01/05/2025 and 30/06/2025. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.
        Terms & Conditions See Full Promo Details
      • Multiple Manufacturers
        Get amazing savings on products for your analyses
        Promo Code: 24135
        Expires2025-07-13
        Days
        26
        Hours
        21
        Minutes
        36
        Seconds
        44
      • Multiple Manufacturers
        Save big on essential products for cancer research
        Promo Code24147
        Expires2025-07-31
      • Multiple Manufacturers
        Enjoy discounts up to 78% on top-quality lab equipment and upgrade your laboratory with the best appliances at the lowest prices.
        Promo Code: 24075
        Expires2025-06-30
        Days
        12
        Hours
        21
        Minutes
        36
        Seconds
        44
      Product Content Correction
      Product Title
      Thermo Scientific™ S1 Pipet Filler Accessories > S1 Accessory, Power Supply with Universal Input; Unit Size: 1 piece