missing translation for 'onlineSavingsMsg'
Learn More
S-Benzoylthiamine O-Monophosphate, MP Biomedicals™ Product Code.: 11498961

S-Benzoylthiamine O-Monophosphate, MP Biomedicals™

Product Code. 11498961 Shop All MP Biomedicals Products
250 mg
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy

Product Code. 11498961

Brand: MP Biomedicals 0215483583

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 22457-89-2
Molecular Formula C19H23N4O6PS
Molecular Weight (g/mol) 466.45
MDL Number MFCD00057343,MFCD00057343
InChI Key BTNNPSLJPBRMLZ-LGMDPLHJSA-N
Synonym benfotiamine, berdi, s-benzoylthiamine o-monophosphate, spectrum5_001042, spectrum1503105, benzoylthiaminmonophosphat, 4-n-4-amino-2-methylpyrimidin-5-yl methyl formamido-3-benzoylsulfanyl pent-3-en-1-yl oxyphosphonic acid, s-e-2-4-amino-2-methylpyrimidin-5-yl methyl-formylamino-5-phosphonooxypent-2-en-3-yl benzenecarbothioateShow MoreShow Less
PubChem CID 5282168
IUPAC Name {[(3Z)-4-{N-[(4-amino-2-methylpyrimidin-5-yl)methyl]formamido}-3-[(Z)-benzoylsulfanyl]pent-3-en-1-yl]oxy}phosphonic acid
SMILES C\C(N(CC1=CN=C(C)N=C1N)C=O)=C(/CCOP(O)(O)=O)SC(=O)C1=CC=CC=C1

Specifications

Color White
Assay Percent Range 99.6%
Quantity 250 mg
Formula Weight 466.47
Physical Form Powder
Chemical Name or Material S-Benzoylthiamine O-Monophosphate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
S-Benzoylthiamine O-Monophosphate, MP Biomedicals™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.