missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-Bay K 8644, Tocris Bioscience™

Product Code. 18767096
Click to view available options
Quantity:
10 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

CaV1.x activator; (S)-(-)-enantiomer of (±)-Bay K 8644

L-type Ca2+-channel activator with positive inotropic, vasoconstrictive and behavioral effects in vivo.

Racemate and (R)-(+)-Enantiomer also available.

Chemical Identifiers

CAS 98625-26-4
Molecular Formula C16H15F3N2O4
Molecular Weight (g/mol) 356.301
InChI Key ZFLWDHHVRRZMEI-ZDUSSCGKSA-N
PubChem CID 6603728
IUPAC Name methyl (4S)-2,6-dimethyl-5-nitro-4-[2-(trifluoromethyl)phenyl]-1,4-dihydropyridine-3-carboxylate
SMILES CC1=C(C(C(=C(N1)C)[N+](=O)[O-])C2=CC=CC=C2C(F)(F)F)C(=O)OC

Specifications

Quantity 10 mg
Synonym s---bay k 8644, bay-k-8644 s---, --s-bayk8644, s---bay k8644, bay-k-8644 s--, methyl 4s-2,6-dimethyl-5-nitro-4-2-trifluoromethyl phenyl-1,4-dihydropyridine-3-carboxylate, s---1,4-dihydro-2,6-dimethyl-5-nitro-4-2-trifluoromethyl phenyl-3-pyridinecarboxylic acid methyl ester, tocris-1544, lopac-b-112, d02rih
Formula Weight 356.3
Percent Purity >99%
Chemical Name or Material (S)-(-)-Bay K 8644
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'