missing translation for 'onlineSavingsMsg'
Learn More
(S)-(-)-1-Boc-2-cyanopyrrolidine, 95%, Thermo Scientific™ Product Code.: 15403698

(S)-(-)-1-Boc-2-cyanopyrrolidine, 95%, Thermo Scientific™

Product Code. p-7038237
250 mg, 250mg
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15403698

Brand: Thermo Scientific Alfa Aesar H57150.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
(S)-(-)-1-Boc-2-pyrrolidinecarbonitrile is used in synthesis of prolyl oligopeptidase inhibitors and is a reagent that is used in the design of potent and selective inhibitors of DPP-4 that exhibits an attractive pharmacokinetic profile as antidiabetics.

Solubility
Slightly soluble in water.

Notes
Moisture and light sensitive. Store away from light, water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store in dark.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 228244-04-0
Molecular Formula C10H16N2O2
Molecular Weight (g/mol) 196.25
MDL Number MFCD01861224
InChI Key MDMSZBHMBCNYNO-UHFFFAOYNA-N
Synonym s-1-boc-2-cyanopyrrolidine, s-tert-butyl 2-cyanopyrrolidine-1-carboxylate, s-1-n-boc-2-cyano-pyrrolidine, tert-butyl 2s-2-cyanopyrrolidine-1-carboxylate, s-1-boc-2-cyano-pyrrolidine, s-1-n-boc-2-cyanopyrrolidine, s-1-boc-2-pyrrolidinecarbonitrile, s-n-boc-pyrrolidine-2-carbonitrile, s---1-boc-2-cyanopyrrolidine, s-n-boc-2-pyrrolidinonitrileShow MoreShow Less
PubChem CID 6951263
IUPAC Name tert-butyl (2S)-2-cyanopyrrolidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCCC1C#N

Specifications

Melting Point 33°C to 36°C
Flash Point 110°C (230°F)
Quantity 250 mg
Sensitivity Light and moisture sensitive
Solubility Information Slightly soluble in water.
Optical Rotation +105° (c=1 in Chloroform)
Formula Weight 196.25
Percent Purity 95%
Chemical Name or Material (S)-(-)-1-Boc-2-cyanopyrrolidine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberQ14H003 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number
Lot NumberA15Z037 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
(S)-(-)-1-Boc-2-cyanopyrrolidine, 95%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.