missing translation for 'onlineSavingsMsg'
Learn More
Rubidium Chloride, Metal min. 99%., MP Biomedicals Product Code.: 11406212

Rubidium Chloride, Metal min. 99%., MP Biomedicals

Product Code. 11406212 Shop All MP Biomedicals Products
50 g
Click to view available options
Quantity:
5 g
10 g
50 g
100 g
This item is not returnable. View return policy

Product Code. 11406212

Brand: MP Biomedicals 0210282550

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Solutions may be slightly hazy

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7791-11-9
Molecular Formula ClRb
Molecular Weight (g/mol) 120.92
MDL Number MFCD00011187
InChI Key FGDZQCVHDSGLHJ-UHFFFAOYSA-M
Synonym rubidium chloride, rubidium chloride rbcl, rubidium monochloride, rbcl, rubidium 1+ chloride, rubidium 1+ ion chloride, unii-n3shc5273s, rubidiumchloride, rubidium-chlorid, rubidium-chlorideShow MoreShow Less
PubChem CID 62683
ChEBI CHEBI:78672
IUPAC Name rubidium(1+) chloride
SMILES [Cl-].[Rb+]

Specifications

Quantity 50 g
Assay Percent Range Metal min. 99%.
Formula Weight 120.9
Chemical Name or Material Rubidium Chloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Rubidium Chloride, Metal min. 99%., MP Biomedicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.