Learn More
Welch Ilmvac™ Rubber Vacuum Hose
Thick-walled red rubber vacuum hose. Welch Ilmvac™ Rubber Vacuum Hose well suited for laboratory vacuum pump processes.
Brand:  Welch Ilmvac™ 828310-4
 View more versions of this product
Product Code. 10061242
  • 322.00 NOK / 1m
Only null left
In Stock
Edge
Add to basket
Description
Description
  • Economical
  • Flexible
  • Resilient
  • Thick-walled
TRUSTED_SUSTAINABILITY
Specifications
Specifications
8 mm
18 mm
1 Each
Red
Rubber
100 cm
8mm ID, 18mm OD
Vacuum Hose
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Welch Ilmvac™ Rubber Vacuum Hose > Diameter (I.D./O.D.): 8mm/18mm