Learn More
Fisherbrand™ Rubber Stopper Cork
Brand:  Fisherbrand™ CB011/S
Product Code. 12336457
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Specifications
20/Pk.
14 mm
Stopper Cork
11 mm
Rubber
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Fisherbrand™ Rubber Stopper Cork > Base Diameter: 11mm