Learn More
Saint Gobain Life Sciences Rubber Gas Sample Bag
Use these bags for gas sample analysis. Saint Gobain Life Sciences Rubber Gas Sample Bags feature a cylindrical shape and an ebonite tube and tap.
Brand: Saint Gobain Life Sciences 5607072
This item is not returnable.
View return policy
Description
- Bags are individually packaged
- Available in packs of ten
- Foil thickness: 5/10mm
- Custom sizes and configurations available on request
Specifications
0.5 L | |
Faucet | |
Rubber | |
Gas Sampling Bag |
Product Content Correction
Product Title
Saint Gobain Life Sciences Rubber Gas Sample Bag > Volme: 0.5L