Learn More
RETSCH Rubber Balls
Brand:  RETSCH 32.354.0001
Product Code. 15254729
  • 660.00 NOK / Pack of 5
Only null left
Estimated Shipment: 01-10-2025
Specifications
Rubber Balls
20 mm
5/Pk.
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
RETSCH Rubber Balls > Diameter: 20mm