Learn More
RPS29 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17374-1-AP-150UL
This item is not returnable.
View return policy
Description
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. RPS29 is a ribosomal protein that is a component of the 40S subunit and a member of the S14P family of ribosomal proteins. The protein, which contains a C2-C2 zinc finger-like domain that can bind to zinc, can enhance the tumor suppressor activity of Ras-related protein 1A (KREV1). It is located in the cytoplasm.Specifications
RPS29 | |
Polyclonal | |
Unconjugated | |
RPS29 | |
40S ribosomal protein S29, ribosomal protein S29, RPS29 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
6235 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P62273 | |
RPS29 | |
RPS29 Fusion Protein Ag11179 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |