missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPL23A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1696.32 NOK - 4796.50 NOK
Specifications
Antigen | RPL23A |
---|---|
Concentration | 0.13 mg/mL |
Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Immunocytochemistry, Immunofluorescence, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16845994
|
Proteintech
16386-1-AP-20UL |
20 μL |
1696.32 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16835994
|
Proteintech
16386-1-AP-150UL |
150 μL |
4796.50 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The function of this protein remains unknown.Specifications
RPL23A | |
Immunohistochemistry (Paraffin), Immunoprecipitation, Immunocytochemistry, Immunofluorescence, Western Blot | |
Unconjugated | |
Rabbit | |
Rat, Mouse, Human | |
P62750, P62751, P62752 | |
268449, 360572, 6147 | |
RPL23A Fusion Protein Ag9424 | |
Primary | |
-20°C | |
RPL23A |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
60S ribosomal protein L23a, MDA20, ribosomal protein L23a, RPL23A | |
Rpl23a | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title