missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPGRIP1L Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA518785
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with canine based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 64,000.
The protein encoded by this gene can localize to the basal body-centrosome complex or to primary cilia and centrosomes in ciliated cells. The encoded protein has been found to interact with nephrocystin-4. Defects in this gene are a cause of Joubert syndrome type 7 and Meckel syndrome type 5. Two transcript variants encoding different isoforms have been found for this gene.Specifications
RPGRIP1L | |
Polyclonal | |
Unconjugated | |
RPGRIP1L | |
1700047E16Rik; 4931437C01; CORS3; DKFZp686C0668; fantom; fantom homolog; Ftm; JBTS7; KIAA1005; MKS5; Nephrocystin-8; NPHP8; PPP1R134; Protein fantom; protein phosphatase 1, regulatory subunit 134; RGD1311099; RPGR-interacting protein 1-like protein; RPGRIP1 like; RPGRIP1L; Rpgrip1-like; RPGRIP1-like protein | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
23322 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q68CZ1 | |
RPGRIP1L | |
Synthetic peptide sequence (DKENNKAKRDILK) corresponding to the internal amino acids of RPGRIP1L | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |