Learn More
Gilac™ Round Dustbin
Round Dustbin is also available with a standard or swing lid.
Brand: Gilac™ G611721-1
405.13 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- 35 L round dustbin.
- Also available with a standard or swing lid.
Specifications
35 L | |
White | |
Not Autoclavable | |
Dust Bin |
9.2 gal. | |
Round | |
HDPE |
Product Content Correction
Product Title
Gilac™ Round Dustbin > Capacity: 35 L, Color: White