Learn More
Lenz Laborglasintrumente Round Bottom Flasks, Narrow Neck
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand: Lenz Laborglasintrumente 07030028
This item is not returnable.
View return policy
Description
- Acc. to DIN ISO 1773 and in addition to standard
- With beaded rim
- Material: DURAN™
Specifications
26 mm | |
95 mm | |
10/Pk. |
50 mL | |
Beaded rim |
Product Content Correction
Product Title
Lenz Laborglasintrumente Round Bottom Flasks, Narrow Neck > 50 mL