Learn More
Eppendorf™ Aerosol-tight Standard Rotor Key
Designed for use with Eppendorf Rotors and Centrifuges
Brand: Eppendorf™ 5810350018
100.00 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
- Centrifuge rotor key for Eppendorf Rotor nut 5810/5810R
Specifications
Rotor Key | |
Rotor nut 5810/5810R | |
Aerosol tight, standard |
Promotions
-
Multiple Manufacturers
Cancer Research Campaign
Save big on essential products for cancer researchPromo Code: 24147Expires: 2025-07-31
-
Multiple Manufacturers
Quality Analysis and Quality Control Testing
Save up to 59% on some of our most popular productsPromo Code: 24140Expires: 2025-08-24
New Offer Save Up to 59% -
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13