Learn More
Hettich™ Rotor Adapter for Carriers no. 1481, 1490 and 1495
Designed for use with Hettich Rotors. Hettich rotors are optimized for continuous duty and long service life
Brand:  Hettich™ 1457
 View more versions of this product
Product Code. 12063874
  • 692.00 NOK / Each
Description
  • Rotor adapter for Hettich carriers
  • For use with Rotofix and Universal centrifuges
  • Universal Centrifuges can be used for both standard and special applications, they are cost-effective and save valuable space in the laboratory
  • ROTOFIX Centrifuges are ideally equipped for tasks in clinical chemistry, it can also play a vital role in cytology
Specifications
Specifications
Rotor Adapter
7 x 1.1 to 1.4 mL
66 mm
7
Round Bottom, Conical, Blood/Urine Collection
8 mm
Hettich Rotors with Carriers no. 1481, 1490 and 1495
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Hettich™ Rotor Adapter for Carriers no. 1481, 1490 and 1495 > 7 x 1.4mL