Learn More
Hettich™ Rotor Adapter for Carriers no. 1481, 1490 and 1495
Designed for use with Hettich Rotors. Hettich rotors are optimized for continuous duty and long service life
Brand:  Hettich™ 1348
 View more versions of this product
Product Code. 10173422
  • 728.00 NOK / Each
Description
  • Rotor adapter for Hettich carriers
  • For use with Rotofix and Universal centrifuges
  • Universal Centrifuges can be used for both standard and special applications, they are cost-effective and save valuable space in the laboratory
  • ROTOFIX Centrifuges are ideally equipped for tasks in clinical chemistry, it can also play a vital role in cytology
Specifications
Specifications
Rotor Adapter
4 x 4 to 10 mL
100 mm
4
Round Bottom
16 mm
Hettich Rotors with Carriers no. 1481, 1490 and 1495
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Hettich™ Rotor Adapter for Carriers no. 1481, 1490 and 1495 > 4 x 10mL