Learn More
Hettich™ Rotor Adapter for Carriers no. 1481, 1490 and 1495
Designed for use with Hettich Rotors. Hettich rotors are optimized for continuous duty and long service life
Brand: Hettich™ 6318
This item is not returnable.
View return policy
Description
- Rotor adapter for Hettich carriers
- For use with Rotofix and Universal centrifuges
- Universal Centrifuges can be used for both standard and special applications, they are cost-effective and save valuable space in the laboratory
- ROTOFIX Centrifuges are ideally equipped for tasks in clinical chemistry, it can also play a vital role in cytology
Specifications
Rotor Adapter | |
50 mL | |
115 mm | |
1 |
Skirted Conical | |
29 mm | |
Carriers no. 1481, 1490 and 1495 | |
1 |
Product Content Correction
Product Title
Hettich™ Rotor Adapter for Carriers no. 1481, 1490 and 1495 > 50mL