Learn More
IKA™ ROTAVISC Vane Spindle
Vane spindle designed for use with IKA™ ROTAVISC Viscometers.
3006.59 NOK - 4597.62 NOK
Specifications
Product Type | Vane Spindle |
---|---|
For Use With (Equipment) | IKA™ ROTAVISC Viscometer |
Material | AISI 316L |
Product Code | Brand | Description | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | ||||||
17277549
|
IKA™
0025006853 |
VAN-SP-1 Vane Spindle |
4597.62 NOK
Each |
Estimated Shipment: 07-08-2025
Log in to see stock. |
||||||
17287549
|
IKA™
0025006854 |
VAN-SP-2 Vane Spindle |
3720.21 NOK
Each |
Estimated Shipment: 07-08-2025
Log in to see stock. |
||||||
17297549
|
IKA™
0025006855 |
VAN-SP-3 Vane Spindle |
3392.65 NOK
Each |
Estimated Shipment: 07-08-2025
Log in to see stock. |
||||||
17207559
|
IKA™
0025006856 |
VAN-SP-4 Vane Spindle |
3182.07 NOK
Each |
Estimated Shipment: 19-06-2025
Log in to see stock. |
||||||
17247569
|
IKA™
0025008376 |
VAN-SP-5 Vane Spindle |
3006.59 NOK
Each |
Estimated Shipment: 04-09-2025
Log in to see stock. |
||||||
Description
- Quick coupling
- Hook hitch
- M2.5 left-handed thread
Specifications
Vane Spindle | |
IKA™ ROTAVISC Viscometer | |
AISI 316L |
Product Title
IKA™ ROTAVISC Vane Spindle