Description
- Variable speed 2 to 25 rpm
- Adjustable inclination
- For tube diameter 17.5, 25 or 32 mm using additional drum rotors (accessory items)
Specifications
Specifications
| Speed Range | 2 to 25 U/min. |
| Voltage | 230 V |
| Depth (Metric) | 21 cm |
| Height (Metric) | 28 cm |
| Length (Metric) | 20 cm |
| Warranty and Service Offering | 15 Months |
Product Content Correction
Product Title
Robin™ Rotary Mixer >