Learn More
Pyrex™ Quickfit™ Rotary Evaporator Splash Head Adapter
Protects against foaming products that may seep into a condenser. Pyrex™ Quickfit™ Rotary Evaporator Splash Head Adapter have an anti-climb mechanism.
1260.00 NOK - 7175.00 NOK
Specifications
Description | Anti-climb |
---|---|
Material | Borosilicate Glass |
For Use With (Equipment) | Rotary evaporators |
Quantity | 1 Each |
Volume (Metric) | 150 mL |
Product Code | Brand | Taper | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Taper | Price | Quantity & Availability | ||||||
12994065
|
Pyrex™
SH15/24 |
Cone: 24/29, Socket: 29/32 |
1490.00 NOK
Each |
Estimated Shipment: 21-07-2025
Log in to see stock. |
||||||
12904075
|
Pyrex™
SH15/29 |
Cone: 29/32, Socket: 29/32 |
1260.00 NOK
Each |
Estimated Shipment: 03-09-2025
Log in to see stock. |
||||||
15577960
|
Pyrex™
SH15/19 |
Cone: 19/26, Socket: 29/32 |
7175.00 NOK
Pack of 5 |
Estimated Shipment: 14-10-2025
Log in to see stock. |
||||||
Description
- For use with rotary evaporators
- Protects against liquid ingress into the condenser 
- Useful for liquids that have tendency to foam 
- Splash heads feature a 29/32 socket
Specifications
Anti-climb | |
Rotary evaporators | |
150 mL |
Borosilicate Glass | |
1 Each |
Product Title
Pyrex™ Quickfit™ Rotary Evaporator Splash Head Adapter