Learn More
Hirschmann Instruments™ rotarus™ Peristaltic Pump Heads PK 10-24
Facilitates quick and simple head changing in a single action without tools. Hirschmann Instruments™ rotarus™ Peristaltic Pump Heads PK 10-24 were developed for rapid handling.
Brand:  Hirschmann Instruments™ 951102021
15465.79 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 15305433
  • 22115.00 NOK / Each
Only null left
Estimated Shipment: 04-08-2025
Description
  • For WS/WT 2.4mm tubes
  • 3 rolls
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Hirschmann Instruments™ rotarus™ Peristaltic Pump Heads PK 10-24 >