missing translation for 'onlineSavingsMsg'
Learn More
Roseoflavin, ≥97%, MP Biomedicals™ Product Code.: 11474591

Roseoflavin, ≥97%, MP Biomedicals™

Product Code. 11474591 Shop All MP Biomedicals Products
10 mg
Click to view available options
Quantity:
10 mg
25 mg
0.1 g
This item is not returnable. View return policy

Product Code. 11474591

Brand: MP Biomedicals 0215425210

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Antibacterial
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51093-55-1
Molecular Formula C18H23N5O6
Molecular Weight (g/mol) 405.411
InChI Key IGQLDUYTWDABFK-GUTXKFCHSA-N
Synonym roseoflavin, roseoflavine, 8-demethyl-8-dimethylamino riboflavin, 1-deoxy-1-8-dimethylamino-7-methyl-2,4-dioxo-3,4-dihydrobenzo g pteridin-10 2h-yl-d-ribitol
PubChem CID 49867612
ChEBI CHEBI:72346
IUPAC Name 8-(dimethylamino)-7-methyl-10-[(2S,3S,4R)-2,3,4,5-tetrahydroxypentyl]benzo[g]pteridine-2,4-dione
SMILES CC1=CC2=C(C=C1N(C)C)N(C3=NC(=O)NC(=O)C3=N2)CC(C(C(CO)O)O)O

Specifications

Color Brown
Assay Percent Range ≥97%
Quantity 10 mg
Formula Weight 405.4
Physical Form Solid
Chemical Name or Material Roseoflavin
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Roseoflavin, ≥97%, MP Biomedicals™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.