Learn More
Brady™ Roll Markers
3750.00 NOK
Specifications
Certifications/Compliance | European Directives 67/548 and 88/379 |
---|---|
Adhesive Type | Permanent Acrylic |
Format | Roll |
Material | Polyester |
Product Type | Pipe Marker |
Product Code | Brand | Display Text | Dimensions (L x W x H) | Height (Metric) Character | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Display Text | Dimensions (L x W x H) | Height (Metric) Character | Price | Quantity & Availability | ||||
11755573
|
Brady™
226384 |
Air | 33 m x 100 mm x 150 mm | 13 mm |
3750.00 NOK
1 roll |
Estimated Shipment: 10-07-2025
Log in to see stock. |
||||
11745573
|
Brady™
226383 |
Air | 33 m x 100 mm x 60 mm | 8 mm |
3750.00 NOK
1 roll |
Estimated Shipment: 10-07-2025
Log in to see stock. |
||||
11775573
|
Brady™
226758 |
Vapeur | 33 m x 100 mm x 60 mm | 8 mm |
3750.00 NOK
1 roll |
Estimated Shipment: 10-07-2025
Log in to see stock. |
||||
11765573
|
Brady™
226613 |
Eau | 33 m x 100 mm x 60 mm | 8 mm |
3750.00 NOK
1 roll |
Estimated Shipment: 10-07-2025
Log in to see stock. |
||||
11735573
|
Brady™
226293 |
Eau incendie | 33 m x 100 mm x 60 mm | 8 mm |
3750.00 NOK
1 roll |
Estimated Shipment: 10-07-2025
Log in to see stock. |
||||
Specifications
European Directives 67/548 and 88/379 | |
Roll | |
Pipe Marker | |
French | |
-40°C to +140°C | |
0.119 mm | |
108.27 ft. | |
3.94 in. |
Permanent Acrylic | |
Polyester | |
Pipe and valve marking | |
-40°F to +284°F | |
0.0047 in. | |
33 m x 100 mm | |
33 m | |
100 mm |
Product Title
Brady™ Roll Markers