Learn More
STEMCELL Technologies RoboSep™ Human Whole Blood and Bone Marrow CD138 Positive Selection Kit II
EasySep™ kit with RoboSep™ buffer and RoboSep™ filter tips
Brand:  STEMCELL Technologies 17887RF
Encompass
Product Code. 17167801
  • 16526.91 NOK / 1 set
Explore more special offers
Description
Description
  • The EasySep™ Human Whole Blood and Bone Marrow CD138 Positive Selection Kit is designed to isolate CD138+ (syndecan-1) cells from whole blood and bone marrow by positive selection
  • Desired cells are targeted with Tetrameric Antibody Complexes recognizing CD138 and dextran-coated magnetic particles
  • The cocktail also contains an antibody to human Fc receptor to minimize nonspecific binding
  • Labeled cells are separated using an EasySep™ magnet without the use of columns
  • Cells of interest remain in the tube while unwanted cells are poured off
  • The CD138 antigen is expressed on normal and malignant plasma cells (but not mature B cells)
  • This product replaces the EasySep™ Human Whole Blood and Bone Marrow CD138 Positive Selection Kit (Catalog #18387) for even faster cell isolations
Specifications
Specifications
Myeloma, Plasma
ELISA
60 mL Whole Blood and Bone Marrow
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
STEMCELL Technologies RoboSep™ Human Whole Blood and Bone Marrow CD138 Positive Selection Kit II