missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RKIP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10575-1-AP-20UL
This item is not returnable.
View return policy
Description
PBP binds ATP, opioids and phosphatidylethanolamine, exhibiting a lower affinity for phosphatidylinositol and phosphatidylcholine. This serine protease inhibitor inhibits thrombin, neuropsin and chymotrypsin but not trypsin, tissue type plasminogen activator and elastase. PBP contains hippocampal cholinergic neurostimulating peptide (HCNP), which may be involved in the function of the presynaptic cholinergic neurons of the central nervous system. HCNP increases the production of choline acetyltransferase but not acetylcholinesterase.Specifications
RKIP | |
Polyclonal | |
Unconjugated | |
PEBP1 | |
HCNP, HCNPpp, Neuropolypeptide h3, PBP, PEBP, PEBP 1, PEBP1, Prostatic binding protein, Raf kinase inhibitor protein, RKIP | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23980, 29542, 5037 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P30086, P31044, P70296 | |
Pebp1 | |
RKIP Fusion Protein Ag0864 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction