missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RIT2 293T Cell Overexpression Lysate (Denatured), Abnova
Brand: Abnova
This item is not returnable.
View return policy
Description
Western Blotting
Specifications
293T | |
RIT2 293T Cell Transient Overexpression Lysate(Denatured) | |
23.98 | |
pCMV-RIT2 full-length | |
100 μL | |
Human |
Western Blot | |
NM_002930 | |
6.8 | |
Transient overexpression cell Lysate was tested with Anti-RIT2 antibody (H00006014-B01) by western blotting | |
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- 100UL RIT2 293T Cell Transient Overexpression Lysa
Signal Word
- Warning
Hazard Category
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
- MIXTURE LIST-Contains : Mercaptoethanol
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction