Learn More
Epredia™ Signature Series™ Pen-Fix™
Achieve fixation and dehydration of specimens simultaneously with Epredia™ Signature Series™ Pen-Fix™, a uniquely buffered alcoholic formalin fixative.
Brand:  Epredia™ 6101
 View more versions of this product
Product Code. 10148438
  • 979.00 NOK / Pack of 4
Description
  • Excellent frozen section fixative
  • Performs well with fatty or thick tissue sections
  • Penetration rate three times faster than 10% Neutral Buffered Formalin
  • Can be used on all open or enclosed tissue processors and in the grossing room
  • Recommended for use in the second and/or third stations of the processor as a secondary fixative
TRUSTED_SUSTAINABILITY
Specifications
Specifications
4 x 1 gal.
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Epredia™ Signature Series™ Pen-Fix™ > 1 Gal. (3.79L) Bottle