missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ribosomal protein L4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1696.32 NOK - 4796.50 NOK
Specifications
Antigen | ribosomal protein L4 |
---|---|
Concentration | 0.18 mg/mL |
Applications | Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunoprecipitation, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16848283
|
Proteintech
11302-1-AP-20UL |
20 μL |
1696.32 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16838283
|
Proteintech
11302-1-AP-150UL |
150 μL |
4796.50 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 60S subunit. The protein belongs to the L4E family of ribosomal proteins. It is located in the cytoplasm. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.Specifications
ribosomal protein L4 | |
Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunoprecipitation, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Mouse, Human, Rat | |
P36578, P50878, Q9D8E6 | |
6124, 64302, 67891 | |
ribosomal protein L4 Fusion Protein Ag1828 | |
Primary | |
-20°C | |
RPL4 |
0.18 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
60S ribosomal protein L1, 60S ribosomal protein L4, ribosomal protein L4, RPL1, RPL4 | |
RPL4 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title