missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rhodium solution for ICP 1g/L HCl matrix, Fisher chemical™

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
4735.00 NOK - 16875.00 NOK
Specifications
Analyte or Component Names | Rh |
---|---|
Concentration | 1000 g/L |
Type | Mass Spectrometry Standard |
Element | Rh |
Solvent or Matrix | HCl 5% |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
12983484
![]() |
Fisher Chemical
S 047 100 |
100 mL | Bottle |
4735.00 NOK
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12993484
![]() |
Fisher Chemical
S 047 250 |
250 mL | Bottle |
9060.00 NOK
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12903494
![]() |
Fisher Chemical
S 047 500 |
500 mL | Bottle |
16875.00 NOK
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Rh | |
Mass Spectrometry Standard | |
HCl 5% |
1000 g/L | |
Rh |
Safety and Handling
H225; H304; H315; H336; H410
P210; P233; P240; P241; P261; P264; P271; P273; P280; P301+P310; P302+P352; P303+P361+P353; P304+P340; P312; P321; P331; P332+P313; P370+P378; P391; P403+P233; P403 +P235; P405; P501
Danger
missing translation for 'shelfLife' : 24 months (12 months after opening)
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Single-element Standard Solution for ICP. Rhodium (Rh) 1000mg/l in HCl 5% (S 047)
Signal Word
- Warning
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- EUH210-Safety data sheet available on request.
Precautionary Statement
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Rhodium solution for ICP 1g/L HCl matrix, Fisher chemical™