Learn More
IKA™ RH Digital White
GREENER_CHOICE
Brand:  IKA™ 0004678000
8477.15 NOK valid until 2025-07-31
Use promo code "24075" to get your promotional price.
Product Code. 15168369
  • 10517.20 NOK / Each
The Ultimate Lab Appliances Promotion
Use promo code 24075 at checkout
View Available Promotions
Only null left
In Stock
Specifications
15 L
White
Digital
Certificates
Promotions
Promotions
Product Content Correction
Product Title
IKA™ RH Digital White > Capacity: 15L