Learn More
RGX-104, MedChemExpress
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
2mg
50mg
5mg
Description
RGX-104 is an orally bioavailable and potent liver-X nuclear hormone receptor (LXR) agonist that modulates innate immunity via transcriptional activation of the ApoE gene.
Specifications
Specifications
Chemical Name or Material | RGX-104 |
CAS | 610318-54-2 |
Color | White |
Purity Grade Notes | Research |
Molecular Formula | C34H33ClF3NO3 |
Quantity | 50 mg |
Solubility Information | DMSO : 100 mg/mL (167.76 mM; Need ultrasonic) |
SMILES | O=C(O)CC1=CC=CC(OCC[C@H](N(CC2=CC=CC(C(F)(F)F)=C2Cl)CC(C3=CC=CC=C3)C4=CC=CC=C4)C)=C1 |
Molecular Weight (g/mol) | 596.08 |
Formula Weight | 596.08 |
Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
RGX-104, MedChemExpress