Learn More
RGX-104, MedChemExpress
MedChemExpress RGX-104 is an orally bioavailable and potent liver-X nuclear hormone receptor (LXR) agonist that modulates innate immunity via transcriptional activation of the ApoE gene.
Brand: Medchem Express HY-111498A
Description
RGX-104 is an orally bioavailable and potent liver-X nuclear hormone receptor (LXR) agonist that modulates innate immunity via transcriptional activation of the ApoE gene.Specifications
RGX-104 | |
White | |
C34H33ClF3NO3 | |
DMSO : 100 mg/mL (167.76 mM; Need ultrasonic) | |
596.08 | |
99.35% | |
Solid |
610318-54-2 | |
Research | |
10 mg | |
O=C(O)CC1=CC=CC(OCC[C@H](N(CC2=CC=CC(C(F)(F)F)=C2Cl)CC(C3=CC=CC=C3)C4=CC=CC=C4)C)=C1 | |
596.08 | |
Research | |
Cancer-programmed cell death |
Product Suggestions
Customers who viewed this item also viewed.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Research purposes only