Learn More
RGX-104, MedChemExpress Product Code.: 16443106
RGX-104, MedChemExpress
Product Code. 16443106
25 mg, 25mg
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
2mg
50mg
5mg
Product Code. 16443106
Brand: Medchem Express HY111498A
MedChemExpress RGX-104 is an orally bioavailable and potent liver-X nuclear hormone receptor (LXR) agonist that modulates innate immunity via transcriptional activation of the ApoE gene.
RGX-104 is an orally bioavailable and potent liver-X nuclear hormone receptor (LXR) agonist that modulates innate immunity via transcriptional activation of the ApoE gene.
Specifications
Chemical Name or Material RGX-104
CAS 610318-54-2
Color White
Purity Grade Notes Research
Molecular Formula C34H33ClF3NO3
Quantity 25 mg
Solubility Information DMSO : 100 mg/mL (167.76 mM; Need ultrasonic)
SMILES O=C(O)CC1=CC=CC(OCC[C@H](N(CC2=CC=CC(C(F)(F)F)=C2Cl)CC(C3=CC=CC=C3)C4=CC=CC=C4)C)=C1
Molecular Weight (g/mol) 596.08
Formula Weight 596.08
Percent Purity 99.35%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
RGX-104, MedChemExpress