Learn More
RGX-104 hydrochloride, MedChemExpress Product Code.: 16460787
RGX-104 hydrochloride, MedChemExpress
Product Code. 16460787
50 mg, 50mg
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
2mg
50mg
5mg
Product Code. 16460787
Brand: Medchem Express HY111498
MedChemExpress RGX-104 hydrochloride is a small-molecule LXR agonist that modulates innate immunity via transcriptional activation of the ApoE gene.
RGX-104 hydrochloride is a small-molecule LXR agonist that modulates innate immunity via transcriptional activation of the ApoE gene.
Specifications
Chemical Name or Material RGX-104 hydrochloride
CAS 610318-03-1
Color White
Purity Grade Notes Research
Molecular Formula C34H34Cl2F3NO3
Quantity 50 mg
Solubility Information DMSO : 150 mg/mL (237.14 mM; ultrasonic and warming and heat to 60°C)
SMILES O=C(O)CC1=CC=CC(OCC[C@H](N(CC2=CC=CC(C(F)(F)F)=C2Cl)CC(C3=CC=CC=C3)C4=CC=CC=C4)C)=C1.Cl
Molecular Weight (g/mol) 632.54
Formula Weight 632.54
Percent Purity 99.14%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
RGX-104 hydrochloride, MedChemExpress