Learn More
Grant Instruments™ Reversible Raised Shelf for Unstirred Water Baths
Allows two shelf depths to accommodates shallow vessels. Grant Instruments™ Reversible Raised Shelf for Unstirred Water Baths can be used with a variety of other Grant Heating Circulators and Heated Circulating Baths.
Brand:  Grant Instruments™ RS28H
 View more versions of this product
Product Code. 11466407
  • 2920.00 NOK / Each
Only null left
Estimated Shipment: 08-09-2025
Description
  • Choose from a variety of sizes to fit your tank.
  • For use with either Stainless Steel and Plastic tank models
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Half the area of SBB Aqua 26 Plus or Sub Aqua 26 Pro
Stainless Steel
45 or 135 mm
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Grant Instruments™ Reversible Raised Shelf for Unstirred Water Baths > Height: 45 or 135mm