Learn More
3M™ Reusable Low Maintenance Half Face Mask Respirator
Reusable Half Face Masks 6000 Series are made from a lightweight, comfortable elastomeric material with a bayonet-style twin-filter design, cradle head harness and easy-fasten neck strap.
3210.00 NOK
Specifications
Color | Gray |
---|---|
Design Type | Bayonet Style |
Disposable | Reusable |
Filter Type | Combination |
Material | Thermoplastic Elastomer |
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
17873246
|
3M™
7100049927 |
Medium |
3210.00 NOK
Pack of 8 |
Estimated Shipment: 25-06-2025
Log in to see stock. |
||||||
17893256
|
3M™
7100049928 |
Large |
3210.00 NOK
Pack of 8 |
Estimated Shipment: 25-06-2025
Log in to see stock. |
||||||
Description
- For economical, low-maintenance reusable masks that are easy to handle, use 3M™ Reusable Half Face Mask 6000 Series.
- Our reusable half masks are designed for simplicity, with an elastomeric material that’s lightweight and comfortable to wear over long shifts, a cradle head harness and easy-fasten neck strap.
- They provide protection against gases, vapours and particulates, and feature a bayonet-style connection that fits to a broad range of twin, lightweight filters, depending on your individual needs.
- The twin-filter design evenly balances weight across the mask, and the low profile means minimal impact to your field of vision and better compatibility with welding and grinding shields.
- They can be used with 3M™ Supplied Air Systems for your increased convenience and flexibility, and connect with all 3M™ 2000, 5000 and 6000 Series filters (except 3M™ 6098 or 6099).
- The 6000 Series has a specially designed range of spares and accessories to help you maintain the right level of comfort and protection when parts become worn or damaged.
- They’re available in three sizes; small, medium and large.
Specifications
Gray | |
Reusable | |
Thermoplastic Elastomer | |
No |
Bayonet Style | |
Combination | |
Reusable Half Facepiece | |
Yes |
Product Title
3M™ Reusable Low Maintenance Half Face Mask Respirator