Learn More
3M™ 7500 Series Reusable Half Facepiece Reusable Respirators
Half-mask respirators with enhanced comfort and visibility use a bayonet attachment system for twin lightweight filters to protect against gases, vapors and particulates.
Brand: 3M™ 7000104177
387.27 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Alert:
These safety products must be used in accordance with OSHA regulations and the user instructions, warnings and limitations accompanying each product. Misuse may result in sickness or death.
Requires : Compatible cartridges and/or filters. Accommodates 3M 7093 and Series 2000 filters and Series 6000 cartridges.
Description
- Durable, reusable and heat-resistant mask is made of advanced silicone material.
- Unique adjustment design helps reduce tension and pressure points on the face.
- Cool Flow™ exhalation valve opens easier, lowering exhalation breathing resistance, reducing heat and moisture buildup.
- Easily adjusted, dual-mode head harness gives option of wearing in traditional or drop-down mode.
- Ideal choice for welding applications.
- Color-coded sizes for easy identification.
- 3M Catalog# 7501, 3M Catalog# 7502, 3M Catalog# 7503
Specifications
EN140 | |
Half Mask | |
K7502 |
Silicone | |
Medium | |
1 Ea. |