Description
- Used with twin lightweight filters attached by a simple bayonet attachment system
- Flexible system (gas/vapor and/or particulate filters plus Supplied-Air option)
- Soft, pliable, hypo-allergenic facepiece material
- Easy to use
- Dual filter design reduces breathing resistance and improves weight distribution for greater comfort
- Large, polycarbonate lens
- Double face sealing gasket ensures a better fit
- Speech diaphragm improves communication
- Six strap harness assembly for easy adjustment and a reliable fit
- Soft compound inner nose-cup increases user comfort
- Spectacle kit available
Specifications
Specifications
Connection Type | Bayonet |
Color | Black |
Product Type | Reusable Full Face Mask Respirator |
Harness Style | 6 strap assembly |
Includes | Polycarbonate visor, six strap head harness, cool-flow valve, silicone rubber facepiece |
Material (Lens) | Polycarbonate |
Quantity | 1 Ea. |
Product Content Correction
Product Title
3M™ Reusable Full Face Respirators >